6-Heptynoic acid 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H53519.03
H53519.06
H53519.14
H53519.03EA
1630
CZK
H53519.03
H53519.06
H53519.14
6-Heptynoic acid 95%
6-Heptynoic acid
Purity: 95%
CAS No.: 30964-00-2
Molecular Formula: C7H10O2
Formula Weight: 126.16
Flash Point: 113°(235°F)
Refractive Index: 1.451
Boiling Point: 93-94°/1mm
Beilstein Reference: 1747024
UN No.: 3265
MDL No.: MFCD00239430
Vzorec:
C₇H₁₀O₂ Bod varu: 93…94 °C (1 mmHg) Hustota: 0,997 Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00239430 CAS číslo: 30964-00-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...