6-Chloro-2-hexanone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H53462.06
H53462.14
H53462.06EA
2370
CZK
H53462.06
H53462.14
6-Chloro-2-hexanone ≥98%
6-Chloro-2-hexanone
Purity: 98%
CAS No.: 10226-30-9
Molecular Formula: C6H11ClO
Formula Weight: 134.61
Flash Point: 91°(196°F)
Refractive Index: 1.4435
Boiling Point: 86°/16mm
MDL No.: MFCD00191625
Vzorec:
C₆H₁₁ClO Bod varu: 86 °C (16 mmHg) Hustota: 1,02 Bod vzplanutí: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191625 CAS číslo: 10226-30-9 EINECS: 233-546-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...