Tisk…

6-Chloro-1-hexyne ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Chlorohex-1-yne

H53476.AA H53476.AC
H53476.AAEA 2290 CZK
H53476.AA H53476.AC
6-Chloro-1-hexyne ≥98%
6-Chloro-1-hexyne

Purity: 98% CAS No.: 10297-06-0 Molecular Formula: C6H9Cl Formula Weight: 116.59 Flash Point: 37°(99°F) Refractive Index: 1.451 Boiling Point: 83°/95mm UN No.: 1993 MDL No.: MFCD00013697

Vzorec: C₆H₉Cl
MW: 116,59 g/mol
Bod varu: 83 °C (95 mmHg)
Hustota: 0,962
Bod vzplanutí: 37 °C (99 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013697
CAS číslo: 10297-06-0
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR