Tisk…

6-Hydroxyflavone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B25016.03 B25016.06
B25016.03EA 1660 CZK
B25016.03 B25016.06
6-Hydroxyflavone ≥98%
6-Hydroxyflavone

Purity: 98% CAS No.: 6665-83-4 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 234-236° Beilstein Reference: 15827

Vzorec: C₁₅H₁₀O₃
Bod varu: 395 °C (1013 hPa)
Bod tání: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00017329
CAS číslo: 6665-83-4
EINECS: 229-704-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR