Tisk…

4-Phenoxybenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

700878-1G 700878-5G
SIAL700878-1GEA 1310 CZK
SIAL700878-1G SIAL700878-5G
4-Phenoxybenzenesulphonyl chloride, Sigma-Aldrich®
4-Phenoxybenzenesulphonyl chloride
Vzorec: C₁₂H₉ClO₃S
Bod varu: 180…190 °C (6 mmHg)
Bod tání: 44 °C
MDL Number: MFCD00625748
CAS číslo: 1623-92-3
UN: 3260
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR