Tisk…

4-Nitrobenzenesulphonamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

120502-25G 120502-5G
SIAL120502-25GEA 6430 CZK
SIAL120502-25G SIAL120502-5G
4-Nitrobenzenesulphonamide, Sigma-Aldrich®
4-Nitrobenzenesulphonamide
Vzorec: C₆H₆N₂O₄S
MW: 202,19 g/mol
Bod tání: 187 °C
MDL Number: MFCD00007937
CAS číslo: 6325-93-5
EINECS: 228-691-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR