4-Methylthiobenzamide ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Methylbenzothioamide, Thio-p-toluamide
B20320.03
B20320.06
B20320.03EA
862
CZK
B20320.03
B20320.06
4-Methylthiobenzamide ≥97%
4-Methylthiobenzamide
Purity: 97%
CAS No.: 2362-62-1
Molecular Formula: C8H9NS
Formula Weight: 151.23
Melting Point: 166-169°
MDL No.: MFCD00173750
Vzorec:
C₈H₉NS MW: 151,23 g/mol Bod tání: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173750 CAS číslo: 2362-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...