Tisk…

4-Iodoimidazole ≥94%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4(5)-Iodoimidazole, 5-Iodo-1H-imidazole, 4-Iodo-1H-imidazole

L20184.03 L20184.06
L20184.03EA 3810 CZK
L20184.03 L20184.06
4-Iodoimidazole ≥94%
4-Iodoimidazole

Purity: 94% CAS No.: 71759-89-2 Molecular Formula: C3H3IN2 Formula Weight: 193.96 Melting Point: 135-138° Beilstein Reference: 1616

Vzorec: C₃H₃IN₂
MW: 193.97
Bod tání: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD01632213
CAS číslo: 71759-89-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR