Tisk…

4-Fluorobenzhydrazide ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Fluorobenzoic hydrazide, 4-Fluorobenzene-1-carbohydrazide, 4-Fluorobenzoic acid hydrazide, 4-Fluorobenzohydrazide

H53496.06 H53496.14
H53496.06EA 2260 CZK
H53496.06 H53496.14
4-Fluorobenzhydrazide ≥97%
4-Fluorobenzhydrazide

Purity: 97% CAS No.: 456-06-4 Molecular Formula: C7H7FN2O Formula Weight: 154.14 Melting Point: 160-162° MDL No.: MFCD00060562

Vzorec: C₇H₇FN₂O
MW: 154,14 g/mol
Bod tání: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00060562
CAS číslo: 456-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR