4-Vinylpyridine 95% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Stabilization:
80 - 150 ppm Stabilised with hydroquinone (HQ)
L13316.AE
L13316.AP
L13316.AC
L13316.AEEA
1570
CZK
L13316.AE
L13316.AP
L13316.AC
4-Vinylpyridine 95% stabilised
4-Vinylpyridine
Stabilized with 100 ppm hydroquinone
Purity: 96%
CAS No.: 100-43-6
Molecular Formula: C7H7N
Formula Weight: 105.14
Storage Temperature: -30°C to -10°C
Flash Point: 51°(124°F)
Refractive Index: 1.5510
Boiling Point: 58-61°/12mm
Beilstein Reference: 104506
UN No.: 3073
MDL No.: MFCD00006447
Vzorec:
C₇H₇N MW: 105.14 Bod varu: 58…61 °C (12 mmHg) Bod tání: –39 °C Hustota: 0.984 Bod vzplanutí: 51 °C (124 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00006447 CAS číslo: 100-43-6 EINECS: 202-852-0 UN: 3073 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...