4-Acetylbenzenesulphonyl chloride 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
343850050
343850500
ACRO343850050EA
1890
CZK
ACRO343850050
ACRO343850500
4-Acetylbenzenesulphonyl chloride 97%
4-Acetylbenzenesulphonyl chloride
Vzorec:
C₈H₇ClO₃S Bod tání: 83…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800269 CAS číslo: 1788-10-9 UN: 3261 ADR: 8,III |
Výsledky specifikačního testu
Appearance | White to orange to brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 83 to 87 °C |
Additional info | product may contain varying amounts of inorganic chloride |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...