4-Acetylbenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Acetylbenzenesulphonyl Chloride, 4-Acetylbenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
00933-10G
00933-50G
SIAL00933-10GEA
3090
CZK
SIAL00933-10G
SIAL00933-50G
4-Acetylbenzenesulphonyl chloride, Sigma-Aldrich®
4-Acetylbenzenesulphonyl chloride
Vzorec:
C₈H₇ClO₃S MW: 218,66 g/mol Bod tání: 87…88 °C |
MDL Number:
MFCD00800269 CAS číslo: 1788-10-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...