4-Chlorobenzenesulphonamide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
C24191-100G
C24191-5G
SIALC24191-100GEA
0
CZK
SIALC24191-100G
SIALC24191-5G
4-Chlorobenzenesulphonamide, Sigma-Aldrich®
4-Chlorobenzenesulphonamide
Vzorec:
C₆H₆ClNO₂S MW: 191,64 g/mol Bod varu: 316 °C (1013 hPa) Bod tání: 144…148 °C |
MDL Number:
MFCD00007936 CAS číslo: 98-64-6 EINECS: 202-689-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...