Tisk…

4-Butylbenzenesulphonyl chloride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4-Butylbenzene-1-sulphonyl chloride

A16577.06 A16577.14
A16577.06EA 2050 CZK
A16577.06 A16577.14
4-Butylbenzenesulphonyl chloride ≥97%
4-Butylbenzenesulphonyl chloride

Purity: 97% CAS No.: 54997-92-1 Molecular Formula: C10H13ClO2S Formula Weight: 232.73 Melting Point: 13-15° Boiling Point: 126-128°/0.6mm UN No.: 3265 MDL No.: MFCD00173908

Vzorec: C₁₀H₁₃ClO₂S
Bod varu: 126…128 °C (0,6 mmHg)
Bod tání: 13…15 °C
Storage Temperature: Ambient
MDL Number: MFCD00173908
CAS číslo: 54997-92-1
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR