Tisk…

4-Bromo-2-methylbenzenesulphonyl chloride 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H31835.03 H31835.09
H31835.03EA 656 CZK
H31835.03 H31835.09
4-Bromo-2-methylbenzenesulphonyl chloride 97%
4-Bromo-2-methylbenzenesulphonyl chloride

Purity: 97% CAS No.: 139937-37-4 Molecular Formula: C7H6BrClO2S Formula Weight: 269.55 Melting Point: 63-65° Boiling Point: 164°/15mm UN No.: 3265 MDL No.: MFCD01365820

Vzorec: C₇H₆BrClO₂S
Bod varu: 164 °C (15 mmHg)
Bod tání: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00219927
CAS číslo: 139937-37-4
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR