Tisk…

4-Aminobenzamidine dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride

M-Clarity™ quality level = MQ200

857661-10G 857661-1G 857661-50G
SIAL857661-10GEA 7210 CZK
SIAL857661-10G SIAL857661-1G SIAL857661-50G
4-Aminobenzamidine dihydrochloride, Sigma-Aldrich®
4-Aminobenzamidine dihydrochloride
Vzorec: C₇H₁₁Cl₂N₃
MW: 208,09 g/mol
Bod tání: 290…291 °C
MDL Number: MFCD00013001
CAS číslo: 2498-50-2
EINECS: 219-692-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR