Tisk…

Biochanin A ≥98%

Dodavatel: Thermo Scientific

Synonyms: 5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-one, 4'-Methoxy-5,7-dihydroxyisoflavone, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-4'-methoxyisoflavone

43853.03 43853.MD 43853.06
43853.03EA 6320 CZK
43853.03 43853.MD 43853.06
Biochanin A ≥98%
Biochanin A

Purity: 98% CAS No.: 491-80-5 Molecular Formula: C16H12O5 Formula Weight: 284.27 Physical Form: Powder Melting Point: 210-213° Beilstein Reference: 278107 MDL No.: MFCD00006839

Vzorec: C₁₆H₁₂O₅
MW: 284,27 g/mol
Bod tání: 210…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00006839
CAS číslo: 491-80-5
EINECS: 207-744-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR