Tisk…

Genistein, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

M-Clarity™ quality level = MQ200

G6649-100MG G6649-25MG G6649-5MG
SIALG6649-100MGEA 12880 CZK
SIALG6649-100MG SIALG6649-25MG SIALG6649-5MG
Genistein, Sigma-Aldrich®
Genistein
Vzorec: C₁₅H₁₀O₅
MW: 270,24 g/mol
Bod tání: 297…298 °C
MDL Number: MFCD00016952
CAS číslo: 446-72-0
EINECS: 207-174-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR