Apigenin ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
L15041.MB
L15041.MC
L15041.MBEA
2200
CZK
L15041.MB
L15041.MC
Apigenin ≥97%
Apigenin
Purity: 97%
CAS No.: 520-36-5
Molecular Formula: C15H10O5
Formula Weight: 270.24
Storage Temperature: 2°C to 8°C
Melting Point: ca 315° dec.
Beilstein Reference: 262620
Merck Reference: 14,730
MDL No.: MFCD00006831
Vzorec:
C₁₅H₁₀O₅ MW: 270,24 g/mol Bod tání: ∼ 315 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006831 CAS číslo: 520-36-5 EINECS: 208-292-3 Merck Index: 14,730 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...