Tisk…

Apigenin

Dodavatel: Cayman Chemical

Synonyms: 4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one

10010275-100 10010275-25 10010275-50 10010275-500
CAYM10010275-100EA 8610 CZK
CAYM10010275-100 CAYM10010275-25 CAYM10010275-50 CAYM10010275-500
Apigenin
Apigenin
Vzorec: C₁₅H₁₀O₅
MW: 270,24 g/mol
MDL Number: MFCD00006831
CAS číslo: 520-36-5
EINECS: 208-292-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR