Tisk…

4,4,5,5,5-Pentafluorpentan-1-ol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4,4,5,5,5-Pentafluoropentanol

M-Clarity™ quality level = MQ100

541133-25G 541133-5G
SIAL541133-25GEA 6970 CZK
SIAL541133-25G SIAL541133-5G
4,4,5,5,5-Pentafluorpentan-1-ol, Sigma-Aldrich®
4,4,5,5,5-Pentafluorpentan-1-ol
Vzorec: C₅H₇F₅O
MDL Number: MFCD00153224
CAS číslo: 148043-73-6
EINECS: 421-360-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR