Tisk…

4,4,5,5,5-Pentafluorpentan-1-ol ≥94%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4,4,5,5,5-Pentafluoropentanol

B21512.04 B21512.09
B21512.04EA 1080 CZK
B21512.04 B21512.09
4,4,5,5,5-Pentafluorpentan-1-ol ≥94%
4,4,5,5,5-Pentafluorpentan-1-ol

Purity: 94% CAS No.: 148043-73-6 Molecular Formula: C5H7F5O Formula Weight: 178.10 Flash Point: 63°(145°F) Refractive Index: 1.3340 Boiling Point: 135° MDL No.: MFCD00153224

Vzorec: C₅H₇F₅O
MW: 178,1 g/mol
Bod varu: 135 °C
Hustota: 1,350
Bod vzplanutí: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00153224
CAS číslo: 148043-73-6
EINECS: 421-360-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR