Tisk…

Ethyl-4,4,4-trifluoroacetoacetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate

M-Clarity™ quality level = MQ100

E50205-100G E50205-25G E50205-5G
SIALE50205-100GEA 9400 CZK
SIALE50205-100G SIALE50205-25G SIALE50205-5G
Ethyl-4,4,4-trifluoroacetoacetate, Sigma-Aldrich®
Ethyl-4,4,4-trifluoroacetoacetate
Vzorec: C₆H₇F₃O₃
MW: 184,11 g/mol
Bod varu: 131 °C (1013 hPa)
Bod tání: –39 °C
Hustota: 1,2586 g/cm³ (15.5 °C)
MDL Number: MFCD00000424
CAS číslo: 372-31-6
EINECS: 206-750-7
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR