Ethyl-4,4,4-trifluoroacetoacetate 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate
Purity: 98+%
CAS No.: 372-31-6
Molecular Formula: C6H7F3O3
Formula Weight: 184.12
Flash Point: 38°(100°F)
Refractive Index: 1.3760
Odour: Odourless
Melting Point: -30°
Boiling Point: 129-130°
Beilstein Reference: 608353
UN No.: 3272
MDL No.: MFCD00000424
Vzorec:
C₆H₇F₃O₃ MW: 184,11 g/mol Bod varu: 129…130 °C Bod tání: –30 °C Hustota: 1,259 Bod vzplanutí: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000424 CAS číslo: 372-31-6 EINECS: 206-750-7 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...