Tisk…

4-Acetoxybenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

A11350.14 A11350.18 A11350.30
A11350.14EA 1400 CZK
A11350.14 A11350.18 A11350.30
4-Acetoxybenzoic acid ≥98%
4-Acetoxybenzoic acid

Purity: 98+% CAS No.: 2345-34-8 Molecular Formula: C9H8O4 Formula Weight: 180.15 Melting Point: 192-195° Beilstein Reference: 1871997

Vzorec: C₉H₈O₄
MW: 180,16 g/mol
Bod tání: 192…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00002540
CAS číslo: 2345-34-8
EINECS: 219-066-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR