4-Acetoxy-3-methoxybenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Formyl-2-methoxyphenyl acetate, Vanillin acetate
M-Clarity™ quality level = MQ100
W310808-100G-K
W310808-1KG-K
W310808-5KG-K
SIALW310808-100G-KEA
2590
CZK
SIALW310808-100G-K
SIALW310808-1KG-K
SIALW310808-5KG-K
4-Acetoxy-3-methoxybenzaldehyde, Sigma-Aldrich®
4-Acetoxy-3-methoxybenzaldehyde
Vzorec:
C₁₀H₁₀O₄ MW: 194,19 g/mol |
MDL Number:
MFCD00003362 CAS číslo: 881-68-5 EINECS: 212-920-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...