Tisk…

4-Acetoxy-3-methoxybenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Formyl-2-methoxyphenyl acetate, Vanillin acetate

M-Clarity™ quality level = MQ100

258601-250G 258601-50G
SIAL258601-250GEA 16740 CZK
SIAL258601-250G SIAL258601-50G
4-Acetoxy-3-methoxybenzaldehyde, Sigma-Aldrich®
4-Acetoxy-3-methoxybenzaldehyde
Vzorec: C₁₀H₁₀O₄
MW: 194,19 g/mol
MDL Number: MFCD00003362
CAS číslo: 881-68-5
EINECS: 212-920-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR