4-(Aminomethyl)benzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
283746-100G
283746-25G
283746-5G
SIAL283746-100GEA
5100
CZK
SIAL283746-100G
SIAL283746-25G
SIAL283746-5G
4-(Aminomethyl)benzoic acid, Sigma-Aldrich®
4-(Aminomethyl)benzoic acid
Vzorec:
C₈H₉NO₂ MW: 151,16 g/mol |
MDL Number:
MFCD00010203 CAS číslo: 56-91-7 EINECS: 200-297-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...