Tisk…

Ethyl-3-phenylpropionate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester

M-Clarity™ quality level = MQ300

W245518-1KG W245518-250G W245518-SAMPLE
SIALW245518-1KGEA 0 CZK
SIALW245518-1KG SIALW245518-250G SIALW245518-SAMPLE
Ethyl-3-phenylpropionate, Sigma-Aldrich®
Ethyl-3-phenylpropionate
Vzorec: C₁₁H₁₄O₂
MW: 178,23 g/mol
Bod varu: 247 °C (1013 hPa)
Hustota: 1,0343 g/cm³ (0 °C)
MDL Number: MFCD00009206
CAS číslo: 2021-28-5
EINECS: 217-966-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR