Tisk…

3-Methyl-1-butyne 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H53439.06 H53439.14
H53439.06EA 3610 CZK
H53439.06 H53439.14
3-Methyl-1-butyne 96%
3-Methyl-1-butyne

Purity: 97% CAS No.: 598-23-2 Molecular Formula: C5H8 Formula Weight: 68.12 Storage Temperature: 2°C to 8°C Flash Point: -30°(-22°F) Refractive Index: 1.374 Boiling Point: 30° UN No.: 3295 MDL No.: MFCD00039853

Vzorec: C₅H₈
Bod varu: 30 °C
Hustota: 0,666
Bod vzplanutí: –30 °C (–22 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00039853
CAS číslo: 598-23-2
EINECS: 209-925-6
UN: 3295
ADR: 3,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR