Tisk…

3-Hydroxy-2-nitro-6-picoline ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

L07074.09 L07074.18
L07074.09EA 3180 CZK
L07074.09 L07074.18
3-Hydroxy-2-nitro-6-picoline ≥99%
3-Hydroxy-2-nitro-6-picoline

Purity: 99% CAS No.: 15128-90-2 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 105-108° Beilstein Reference: 974751

Vzorec: C₆H₆N₂O₃
Bod tání: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006259
CAS číslo: 15128-90-2
EINECS: 239-192-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR