5-Fluoro-2-methylbenzenesulphonyl chloride ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
5-Fluoro-2-methylbenzene-1-sulphonyl chloride, 3-Fluor-6-methylbenzenesulphonyl chloride
A12641.06
A12641.03
A12641.06EA
4190
CZK
A12641.06
A12641.03
5-Fluoro-2-methylbenzenesulphonyl chloride ≥97%
5-Fluoro-2-methylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 445-05-6
Molecular Formula: C7H6ClFO2S
Formula Weight: 208.64
Flash Point:
Vzorec:
C₇H₆ClFO₂S Bod varu: 130…132 °C (15 mmHg) Hustota: 1,43 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052958 CAS číslo: 445-05-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...