5-Fluoro-o-anisaldehyde ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Fluoro-2-anisaldehyde, 5-Fluoro-2-methoxybenzaldehyde
B24684.03
B24684.06
B24684.14
B24684.03EA
639
CZK
B24684.03
B24684.06
B24684.14
5-Fluoro-o-anisaldehyde ≥98%
5-Fluoro-o-anisaldehyde
Purity: 98%
CAS No.: 19415-51-1
Molecular Formula: C8H7FO2
Formula Weight: 154.14
MDL No.: MFCD00143458
Vzorec:
C₈H₇FO₂ MW: 154,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00143458 CAS číslo: 19415-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...