3-Cyano-4-methylpyridine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile
L19769.03
L19769.06
L19769.14
L19769.03EA
1300
CZK
L19769.03
L19769.06
L19769.14
3-Cyano-4-methylpyridine ≥97%
3-Cyano-4-methylpyridine
Purity: 97%
CAS No.: 5444-01-9
Molecular Formula: C7H6N2
Formula Weight: 118.14
Odour: Characteristic
Melting Point: 41-44°
Boiling Point: 108-111°/20mm
Beilstein Reference: 112016
MDL No.: MFCD00234272
Vzorec:
C₇H₆N₂ MW: 118.14 Bod varu: 108…111 °C (20 mmHg) Bod tání: 41…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234272 CAS číslo: 5444-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...