3-Chlorobenzenesulphonamide ≥98%
Dodavatel: Thermo Scientific
L08349.03
L08349.06
L08349.03EA
1690
CZK
L08349.03
L08349.06
3-Chlorobenzenesulphonamide ≥98%
3-Chlorobenzenesulphonamide
Purity: 98%
CAS No.: 17260-71-8
Molecular Formula: C6H6ClNO2S
Formula Weight: 191.64
Melting Point: 144-148°
Beilstein Reference: 2832245
Vzorec:
C₆H₆ClNO₂S MW: 191,64 g/mol Bod tání: 144…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051976 CAS číslo: 17260-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...