5-Chloro-2-methylbenzenesulphonyl chloride ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H32912.06
H32912.03
H32912.06EA
9340
CZK
H32912.06
H32912.03
5-Chloro-2-methylbenzenesulphonyl chloride ≥97%
5-Chloro-2-methylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 34981-38-9
Molecular Formula: C7H6Cl2O2S
Formula Weight: 225.09
Refractive Index: 1.5697
Vzorec:
C₇H₆Cl₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD09742738 CAS číslo: 34981-38-9 EINECS: 252-306-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...