Tisk…

3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L19711.03 L19711.06
L19711.03EA 1920 CZK
L19711.03 L19711.06
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride ≥97%
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride

Purity: 97% CAS No.: 351003-46-8 Molecular Formula: C7H3BrClF3O2S Formula Weight: 323.52 Flash Point: 66°(150°F) Refractive Index: 1.5240 Boiling Point: 217-218° UN No.: 3265 MDL No.: MFCD03094393

Vzorec: C₇H₃BrClF₃O₂S
Bod varu: 217…218 °C
Hustota: 1,835
Bod vzplanutí: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD03094393
CAS číslo: 351003-46-8
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR