3-Bromo-5-nitrosalicylaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Bromo-2-hydroxy-5-nitrobenzaldehyde
M-Clarity™ quality level = MQ100
652784-1G
652784-5G
SIAL652784-1GEA
1450
CZK
SIAL652784-1G
SIAL652784-5G
3-Bromo-5-nitrosalicylaldehyde, Sigma-Aldrich®
3-Bromo-5-nitrosalicylaldehyde
Vzorec:
C₇H₄BrNO₄ MW: 246,02 g/mol |
MDL Number:
MFCD00051833 CAS číslo: 16789-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...