Tisk…

3-Amino-2-naphthol 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-Amino-2-hydroxynaphthalene, 3-Aminonaphthalen-2-ol

B24570.03 B24570.06 B24570.14
B24570.03EA 1880 CZK
B24570.03 B24570.06 B24570.14
3-Amino-2-naphthol 97%
3-Amino-2-naphthol

Purity: 97% CAS No.: 5417-63-0 Molecular Formula: C10H9NO Formula Weight: 159.19 Odour: Odourless Melting Point: 232-238° MDL No.: MFCD00004113

Vzorec: C₁₀H₉NO
MW: 159,19 g/mol
Bod tání: 232…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00004113
CAS číslo: 5417-63-0
EINECS: 226-519-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR