3-Amino-2,6-dichloropyridine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine
H31598.06
H31598.14
H31598.06EA
2130
CZK
H31598.06
H31598.14
3-Amino-2,6-dichloropyridine 97%
3-Amino-2,6-dichloropyridine
Purity: 97%
CAS No.: 62476-56-6
Molecular Formula: C5H4Cl2N2
Formula Weight: 163.01
Melting Point: 120-124°
MDL No.: MFCD00023417
Vzorec:
C₅H₄Cl₂N₂ MW: 163,01 g/mol Bod varu: 305 °C (1013 hPa) Bod tání: 120…124 °C Hustota: 1,497 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00023417 CAS číslo: 62476-56-6 EINECS: 263-559-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...