3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1,2-Dimethoxy-1-cyclobutene-3,4-dione, Dimethyl squarate, Squaric acid dimethylester
A18186.06
A18186.03
A18186.06EA
5940
CZK
A18186.06
A18186.03
3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥98%
3,4-Dimethoxy-3-cyclobutene-1,2-dione
Purity: 98%
CAS No.: 5222-73-1
Molecular Formula: C6H6O4
Formula Weight: 142.10
Storage Temperature: 2°C to 8°C
Melting Point: 53-56°
MDL No.: MFCD00101316
Vzorec:
C₆H₆O₄ Bod varu: 247 °C (1013 hPa) Bod tání: 53…56 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00101316 CAS číslo: 5222-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...