3-(Methylthio)-1-propanol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-(Methylthio)propan-1-ol
A10403.06
A10403.14
A10403.22
A10403.06EA
1720
CZK
A10403.06
A10403.14
A10403.22
3-(Methylthio)-1-propanol ≥98%
3-(Methylthio)-1-propanol
Purity: 98%
CAS No.: 505-10-2
Molecular Formula: CH3S(CH2)3OH
Formula Weight: 106.19
Flash Point: 90°(195°F)
Refractive Index: 1.4904
Boiling Point: 89-90°/13mm
MDL No.: MFCD00036560
Vzorec:
C₄H₁₀OS MW: 106.19 Bod varu: 89…90 °C (13 mmHg) Hustota: 1.027 Bod vzplanutí: 90 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036560 CAS číslo: 505-10-2 EINECS: 208-004-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...