Tisk…

Naphthol AS-D chloroacetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthyl chloroacetate

M-Clarity™ quality level = MQ200

N0758-100MG N0758-1G N0758-250MG N0758-500MG
SIALN0758-100MGEA 3190 CZK
SIALN0758-100MG SIALN0758-1G SIALN0758-250MG SIALN0758-500MG
Naphthol AS-D chloroacetate, Sigma-Aldrich®
Naphthol AS-D chloroacetate
Vzorec: C₂₀H₁₆ClNO₃
MW: 353,8 g/mol
CAS číslo: 35245-26-2
EINECS: 252-463-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR