Tisk…

Flavone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Phenylchromen-4-one

F2003-1G F2003-5G
SIALF2003-1GEA 1230 CZK
SIALF2003-1G SIALF2003-5G
Flavone, Sigma-Aldrich®
Flavone
Vzorec: C₁₅H₁₀O₂
MW: 222.24
Bod varu: 180…185 °C (1 mmHg)
Bod tání: 96
MDL Number: MFCD00006825
CAS číslo: 525-82-6
EINECS: 208-383-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR