Flavone 99+%
Dodavatel: Thermo Fisher Scientific
Synonyms:
2-Phenylchromen-4-one
119160010
119160050
119160250
ACRO119160010EA
1010
CZK
ACRO119160010
ACRO119160050
ACRO119160250
Flavone 99+%
Flavone
Vzorec:
C₁₅H₁₀O₂ MW: 222.24 Bod tání: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006825 CAS číslo: 525-82-6 EINECS: 208-383-8 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 95 to 98 °C |
HPLC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...