Tisk…

β-Naphthoflavone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5,6-Benzoflavone, 3-Phenyl-1H-naphtho[2,1-b]pyran-1-one, 3-Phenyl-1H-benzo[f]chromen-1-one

N3633-1G N3633-5G
SIALN3633-1GEA 1520 CZK
SIALN3633-1G SIALN3633-5G
β-Naphthoflavone, Sigma-Aldrich®
β-Naphthoflavone
Vzorec: C₁₉H₁₂O₂
MW: 272,3 g/mol
Bod varu: 461 °C (1013 hPa)
Bod tání: 163…164 °C
Hustota: 1,11 g/cm³ (20 °C)
MDL Number: MFCD00004986
CAS číslo: 6051-87-2
EINECS: 227-958-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR