Tisk…

α-Naphthoflavone ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-Phenyl-4H-benzo[h]chromen-4-one, 2-Phenylbenzo[h]chromen-4-one, 7,8-Benzoflavone

A18542.06 A18542.14
A18542.06EA 2200 CZK
A18542.06 A18542.14
α-Naphthoflavone ≥97%
α-Naphthoflavone

Purity: 97% CAS No.: 604-59-1 Molecular Formula: C19H12O2 Formula Weight: 272.31 Odour: Odourless Melting Point: 156-159° Beilstein Reference: 210862 MDL No.: MFCD00004985

Vzorec: C₁₉H₁₂O₂
Bod varu: 431 °C (1013 hPa)
Bod tání: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00004985
CAS číslo: 604-59-1
EINECS: 210-071-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR