Tisk…

Galangin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,5,7-Trihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 3,5,7-Trihydroxy-2-phenyl-4-benzopyrone, Norizalpinin, 3,5,7-Trihydroxyflavone, 3,5,7-Trihydroxy-2-phenylchromen-4-one, 3,5,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 3,5,7-Trihydroxy-2-phenyl-4H-1-benzopyran-4-one

282200-100MG 282200-25MG
SIAL282200-100MGEA 18330 CZK
SIAL282200-100MG SIAL282200-25MG
Galangin, Sigma-Aldrich®
Galangin
Vzorec: C₁₅H₁₀O₅
MDL Number: MFCD00006833
CAS číslo: 548-83-4
EINECS: 208-960-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR