Tisk…

2-Nitrobenzenesulphonamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

228907-25G 228907-5G
SIAL228907-25GEA 5890 CZK
SIAL228907-25G SIAL228907-5G
2-Nitrobenzenesulphonamide, Sigma-Aldrich®
2-Nitrobenzenesulphonamide
Vzorec: C₆H₆N₂O₄S
MW: 202,19 g/mol
Bod tání: 195…196,5 °C
MDL Number: MFCD00009807
CAS číslo: 5455-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR