2-Methyl-5-nitrobenzenesulphonyl chloride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Nitrotoluene-2-sulphonyl chloride, 2-Methyl-5-nitrobenzene-1-sulphonyl chloride
A14760.14
A14760.06
A14760.14EA
6760
CZK
A14760.14
A14760.06
2-Methyl-5-nitrobenzenesulphonyl chloride ≥97%
2-Methyl-5-nitrobenzenesulphonyl chloride
Purity: 97%
CAS No.: 121-02-8
Molecular Formula: C7H6ClNO4S
Formula Weight: 235.65
Flash Point:
Vzorec:
C₇H₆ClNO₄S MW: 235,65 g/mol Bod varu: 135…137 °C (0,7 mmHg) Bod tání: 42…46 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051695 CAS číslo: 121-02-8 EINECS: 204-444-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...